Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SC 144 hydrochloride
GREENER_CHOICESDP

Catalog No. 496310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-631-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 49-631-0 Supplier Tocris Bioscience™ Supplier No. 4963/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

gp130 inhibitor; blocks cytokine-triggered gp130 signaling

SC 144 hydrochloride is an inhibitor of gp130 (IC50 = 0.72 μM); blocks cytokine-triggered gp130 signaling. SC 144 hydrochloride induces gp130 phosphorylation and down regulates gp130 glycosylation; also abrogates STAT3 phosphorylation, nuclear translocation and inhibits the expression of downstream target genes. SC 144 hydrochloride delays tumor growth in a mouse xenograft model of human ovarian cancer. Orally active.

Specifications

Quantity 10 mg
Product Type SC 144 hydrochloride
Purity >99%
Molecular Weight (g/mol) 358.76
Inhibitors SC 144 hydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.