Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SC 19220
GREENER_CHOICE

Catalog No. 120610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-061-0 10 mg
12-065-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-061-0 Supplier Tocris Bioscience™ Supplier No. 1206/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective EP1 receptor antagonist

Selective EP1 receptor antagonist (IC50 = 6.7 μM for inhibition of [3H]-PGE2 binding to EP1 transfected COS cells).

Chemical Identifiers

CAS 19395-87-0
Molecular Formula C16H14ClN3O3

Specifications

Quantity 10 mg
Formula Weight 331.76
Purity >99%
Molecular Weight (g/mol) 331.756
Chemical Name or Material SC 19220
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.