Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SC 236
GREENER_CHOICESDP

Catalog No. 391910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-191-0 10 mg
39-195-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-191-0 Supplier Tocris Bioscience™ Supplier No. 3919/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective cyclooxygenase-2 (COX-2) inhibitor

Selective COX-2 inhibitor (IC50 values are 0.005 and 17.8 μM for COX-2 and COX-1 respectively). Displays anti-inflammatory properties and potent antimetastatic activity against both experimental metastases and spontaneous metastases arising following primary tumor excision.

Chemical Identifiers

CAS 170569-86-5
Molecular Formula C16H11ClF3N3O2S

Specifications

Quantity 10 mg
Formula Weight 401.79
Purity >99%
Molecular Weight (g/mol) 401.788
Chemical Name or Material SC 236
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.