Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SC 26196
GREENER_CHOICESDP

Catalog No. 418950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-895-0 50 mg
41-891-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-895-0 Supplier R&D Systems™ Supplier No. 4189/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective fatty acid desaturase 2 inhibitor

Selective fatty acid desaturase 2 (FADS2, also known as Δ6 desaturase) inhibitor (IC50 = 0.2 μM in vitro). Displays selectivity over FADS1 and stearoly-CoA 9-desaturase (Δ5 desaturase and SCD-1/Δ9 desaturase, respectively) (IC50 values are >200 μM in vitro). Exhibits anti-inflammatory properties in a mouse edema model. Eliminates CSCs from ovarian cancer cell lines and inhibits sphere formation in vitro.

Chemical Identifiers

CAS 218136-59-5
Molecular Formula C27H29N5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 423.55
Purity >98%
Molecular Weight (g/mol) 423.564
Chemical Name or Material SC 26196
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.