Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SC 51322

Catalog No. 279110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-911-0 10 mg
27-915-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-911-0 Supplier Tocris Bioscience™ Supplier No. 2791/10
This item is not returnable. View return policy

Potent EP1 receptor antagonist

Potent EP1 prostanoid receptor antagonist (Ki = 13.8 nM). Displays analgesic properties in vivo.

Chemical Identifiers

CAS 146032-79-3
Molecular Formula C22H20ClN3O4S

Specifications

Quantity 10 mg
Formula Weight 457.93
Purity >98%
Molecular Weight (g/mol) 457.929
Chemical Name or Material SC 51322
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.