Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SC 514, Tocris Bioscience™

Catalog No. p-4784512
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

IKKβ inhibitor; attenuates NF-κB-induced gene expression

Orally active, ATP-competitive IKKβ inhibitor (IC50 = 3 - 12 μM) that displays > 10-fold selectivity over 28 other kinases including JNK, p38, MK2 and ERK. Attenuates NF-κB-induced gene expression of IL-6, IL-8 and COX-2 in synovial fibroblasts (IC50 values are 20, 20 and 8 μM respectively). Reduces iNOS induction and exhibits anti-inflammatory activity in vivo.

Chemical Identifiers

CAS 354812-17-2
Molecular Formula C9H8N2OS2
Molecular Weight (g/mol) 224.296
InChI Key BMUACLADCKCNKZ-UHFFFAOYSA-N
Synonym 3-amino-5-thiophen-3-yl thiophene-2-carboxamide, 4-amino-2,3'-bithiophene-5-carboxamide, ikk-2 inhibitor, sc-514, insolution ikk-2 inhibitor, sc-514, 5-thien-3-yl-3-aminothiophene-2-carboxamide, curator_000007, 4-amino-2,3-bithiophene-5-carboxamide, biomolki_000076, biomolki2_000080, d0t6va
PubChem CID 2807869
IUPAC Name 3-amino-5-thiophen-3-ylthiophene-2-carboxamide
SMILES C1=CSC=C1C2=CC(=C(S2)C(=O)N)N

Specifications

Quantity 10 mg
Formula Weight 224.3
Percent Purity >98%
Chemical Name or Material SC 514
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.