Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SC 79
GSA_VAGREENER_CHOICE

Catalog No. 463510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-351-0 10 mg
46-355-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-351-0 Supplier Tocris Bioscience™ Supplier No. 4635/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Akt activator

Activator of Akt; binds to the pleckstrin homology domain of Akt. Enhances Akt phosphorylation by upstream protein kinases; also enables cytosolic activation of Akt. Shown to suppress excitotoxicity-induced neuronal death in vitro and in vivo. Exhibits brain penetrance.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 305834-79-1
Molecular Formula C17H17ClN2O5

Specifications

Quantity 10 mg
Formula Weight 364.78
Purity >99%
Molecular Weight (g/mol) 364.782
Chemical Name or Material SC 79
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.