Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ SC-Lys
Click to view available options
Quantity:
30 g
Description
- Single dropout formulation of Synthetic Complete Supplement Mixture (SC) of amino acids for S. cerevisiae growth media.
- Also known as Hopkins Mixture and is available in complete and dropout formulations.

Specifications
Specifications
Content And Storage | Store at Room Temperature. |
Product Type | Cell Culture Media |
Quantity | 30 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction