Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SC4MOL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548651
Description
SC4MOL Polyclonal Antibody for Western Blot, IHC (P)
Sterol-C4-mehtyl oxidase-like protein was isolated based on its similarity to the yeast ERG25 protein. It contains a set of putative metal binding motifs with similarity to that seen in a family of membrane desaturases-hydroxylases.Sterol-C4-mehtyl oxidase-like protein was isolated based on its similarity to the yeast ERG25 protein. It contains a set of putative metal binding motifs with similarity to that seen in a family of membrane desaturases-hydroxylases. The protein is localized to the endoplasmic reticulum membrane and is believed to function in cholesterol biosynthesis. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| SC4MOL | |
| Polyclonal | |
| Unconjugated | |
| MSMO1 | |
| 1500001G16Rik; C-4 methylsterol oxidase; C-4 methylsterol oxidase-like protein; C78600; DESP4; ERG25; MCCPD; Methylsterol monooxygenase 1; methylsterol monoxygenase 1; MGC104344; MSMO1; Neuropep 1; neurorep 1; RANP-1; sc4mol; sterol-C4-methyl oxidase; sterol-C4-methyl oxidase-like; sterol-C4-methyl oxidase-like protein; wu:fb59g06; wu:fb66e06; zgc:56437 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 6307 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q15800 | |
| MSMO1 | |
| KLH conjugated synthetic peptide between 264-293 amino acids from the C-terminal region of human ERG25. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction