Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SC4MOL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563250
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563250 Supplier Invitrogen™ Supplier No. PA563250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATNESVSIF SSASLAVEYV DSLLPENPLQ EPFKNAWNYM LNN Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 84%.

Sterol-C4-mehtyl oxidase-like protein was isolated based on its similarity to the yeast ERG25 protein. It contains a set of putative metal binding motifs with similarity to that seen in a family of membrane desaturases-hydroxylases.Sterol-C4-mehtyl oxidase-like protein was isolated based on its similarity to the yeast ERG25 protein. It contains a set of putative metal binding motifs with similarity to that seen in a family of membrane desaturases-hydroxylases. The protein is localized to the endoplasmic reticulum membrane and is believed to function in cholesterol biosynthesis. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SC4MOL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MSMO1
Gene Accession No. Q15800
Gene Alias 1500001G16Rik; C-4 methylsterol oxidase; C-4 methylsterol oxidase-like protein; C78600; DESP4; ERG25; MCCPD; Methylsterol monooxygenase 1; methylsterol monoxygenase 1; MGC104344; MSMO1; Neuropep 1; neurorep 1; RANP-1; sc4mol; sterol-C4-methyl oxidase; sterol-C4-methyl oxidase-like; sterol-C4-methyl oxidase-like protein; wu:fb59g06; wu:fb66e06; zgc:56437
Gene Symbols MSMO1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SC4MOL. Recombinant protein control fragment (Product #RP-101106).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6307
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.