Learn More
Invitrogen™ SC65 Polyclonal Antibody

Description
Immunogen sequence: FHRARWGLEE EDFQPREEAM LYHNQTAELR ELLEFTHMYL QSDDEMELEE TEPPLEPEDA LSDAEFEGEG DYEEGMYADW WQEPDAKGDE AEAEPEPELA; Positive Samples: U-87MG; Cellular Location: Chromosome, Nucleus, nucleolus.
Specifications
Specifications
| Antigen | SC65 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.89 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | P3H4 |
| Gene Accession No. | Q64375, Q92791 |
| Gene Alias | 1110036O03Rik; AI413214; Endoplasmic reticulum protein SC65; leprecan-like 4; leprecan-like protein 4; Leprel4; NO55; NOL55; nucleolar autoantigen (55kD); nucleolar autoantigen No55; nucleolar autoantigen, 55kDa; P3H4; Prolyl 3-hydroxylase family member 4; prolyl 3-hydroxylase family member 4 (non-enzymatic); SC65; SC65 synaptonemal complex protein; SC65 synaptonemal complex protein (Sc65-pending); synaptonemal complex protein SC65 |
| Gene Symbols | P3H4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.