Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SC65 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989833
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9833 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9833 Supplier Bioss Supplier No. BS17258R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This nucleolar protein was first characterized because it was an autoantigen in cases on interstitial cystitis. The protein, with a predicted molecular weight of 50 kDa, appears to be localized in the particulate compartment of the interphase nucleolus, with a distribution distinct from that of nucleolar protein B23. During mitosis it is associated with chromosomes.

Specifications

Antigen SC65
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene P3H4
Gene Accession No. Q92791
Gene Alias 1110036O03Rik; AI413214; Endoplasmic reticulum protein SC65; leprecan-like 4; Leprecan-like protein 4; LEPREL4; NO55; NOL55; nucleolar autoantigen (55kD); Nucleolar autoantigen No55; nucleolar autoantigen, 55kDa; P3h4; Prolyl 3-hydroxylase family member 4; prolyl 3-hydroxylase family member 4 (non-enzymatic); SC65; SC65 synaptonemal complex protein; SC65 synaptonemal complex protein (Sc65-pending); Synaptonemal complex protein SC65
Gene Symbols P3H4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SC65.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10609
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.