Learn More
Invitrogen™ Sca-1 Monoclonal Antibody (CT-6A/6E)

Description
MA5-17833 targets Sca-1/Ly-6A.2 in FACS applications and shows reactivity with Mouse samples. The MA5-17833 immunogen is mouse Sca-1.
Specifications
Specifications
| Antigen | Sca-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | CT-6A/6E |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Ly6a |
| Gene Accession No. | P05533, Q64253 |
| Gene Alias | 9804; locus A/E; Locus AE; Ly6; Ly67; Ly6a; Ly-6A.2; Ly-6A.2/Ly-6E.1; Ly6A/E; Ly-6A/E; ly6a-e; Ly6e; Ly-6E; Ly-6E.1; lymphocyte antigen 6 complex; lymphocyte antigen 6 complex, locus A; lymphocyte antigen 6 complex, locus E; lymphocyte antigen 6A-2/6E-1; Lymphocyte antigen 6E; RIG-E; Sca1; Sca-1; Sca-2; stem cell antigen 1; Stem cell antigen 2; TAP; T-cell-activating protein; thymic shared antigen 1; Tsa1; TSA-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.