Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCAF11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565407
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565407 Supplier Invitrogen™ Supplier No. PA565407
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NVESVVNEEKI TESSLVEITE HKDFTLKTEE LIESPKLESS EGEIIQTVDR QSVKSPEVQL LGHVETEDVE IIATCDTFGN ED Highest antigen sequence identity to the following orthologs - mouse 42%, rat 37%.

Plays a role in pre-mRNA alternative splicing by regulating spliceosome assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCAF11
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCAF11
Gene Accession No. Q99590
Gene Alias 1110061H03Rik; 17D9; 2610510E10Rik; AI462454; CASP11; Caspase11; Caspase-11; CTD-associated SR protein 11; mKIAA3013; p30 subunit; protein SCAF11; Renal carcinoma antigen NY-REN-40; SC35-interacting protein 1; Scaf11; serine/arginine-rich splicing factor 2, interacting protein; serine/arginine-rich splicing factor 2-interacting protein; SFRS2-interacting protein; SFRS2IP; SIP1; splicing factor, arginine/serine-rich 2, interacting protein; splicing factor, arginine/serine-rich 2-interacting protein; Splicing regulatory protein 129; SR-related and CTD-associated factor 11; SR-related CTD associated factor 11; SR-related CTD-associated factor 11; SRrp129; SRSF2-interacting protein; SRSF2IP
Gene Symbols SCAF11
Host Species Rabbit
Immunogen Recombinant Human SCAF11. Recombinant protein control fragment (Product #RP-106089).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9169
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.