Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCAF11 Polyclonal Antibody

Catalog No. PIPA5121625
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121625 20 μL
PIPA5119371 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121625 Supplier Invitrogen™ Supplier No. PA5121625
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SCAF11 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Plays a role in pre-mRNA alternative splicing by regulating spliceosome assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCAF11
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SCAF11
Gene Accession No. 0
Gene Alias 1110061H03Rik; 17D9; 2610510E10Rik; AI462454; CASP11; Caspase11; Caspase-11; CTD-associated SR protein 11; mKIAA3013; p30 subunit; protein SCAF11; Renal carcinoma antigen NY-REN-40; SC35-interacting protein 1; Scaf11; serine/arginine-rich splicing factor 2, interacting protein; serine/arginine-rich splicing factor 2-interacting protein; SFRS2-interacting protein; SFRS2IP; SIP1; splicing factor, arginine/serine-rich 2, interacting protein; splicing factor, arginine/serine-rich 2-interacting protein; Splicing regulatory protein 129; SR-related and CTD-associated factor 11; SR-related CTD associated factor 11; SR-related CTD-associated factor 11; SRrp129; SRSF2-interacting protein; SRSF2IP
Gene Symbols SCAF11
Host Species Rabbit
Immunogen Recombinant protein Protein SCAF11.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72193
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.