Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCAF11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565434
Description
Immunogen sequence: SFKFVEQQSYK RKSEQEFSFD TPADRSGWTS ASSWAVRKTL PADVQNYYSR RGRNSSGPQS GWMKQEEETS GQDSSLKDQT NQ Highest antigen sequence identity to the following orthologs - mouse 86%, rat 87%.
Plays a role in pre-mRNA alternative splicing by regulating spliceosome assembly.
Specifications
SCAF11 | |
Polyclonal | |
Unconjugated | |
SCAF11 | |
1110061H03Rik; 17D9; 2610510E10Rik; AI462454; CASP11; Caspase11; Caspase-11; CTD-associated SR protein 11; mKIAA3013; p30 subunit; protein SCAF11; Renal carcinoma antigen NY-REN-40; SC35-interacting protein 1; Scaf11; serine/arginine-rich splicing factor 2, interacting protein; serine/arginine-rich splicing factor 2-interacting protein; SFRS2-interacting protein; SFRS2IP; SIP1; splicing factor, arginine/serine-rich 2, interacting protein; splicing factor, arginine/serine-rich 2-interacting protein; Splicing regulatory protein 129; SR-related and CTD-associated factor 11; SR-related CTD associated factor 11; SR-related CTD-associated factor 11; SRrp129; SRSF2-interacting protein; SRSF2IP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9169 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99590 | |
SCAF11 | |
Recombinant Human SCAF11. Recombinant protein control fragment (Product #RP-106114). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction