Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCAF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557334
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557334 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557334 Supplier Invitrogen™ Supplier No. PA557334
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSMTPETVKD VGFGSLVIPG GSVASNLATS ALPAGNVFNA PTKQAEPEEK VPHLIDHQIS SGENTRSVIP NDISSNAAIL G Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.

Regulates the p53 pathway to control the expansion growth of digestive organs. Target protein binds the repetitive C-terminal domain (CTD) of RNA polymerase II; may act with other CTD binding proteins to link transcription and pre-mRNA processing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCAF8
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Scaf8
Gene Accession No. Q9UPN6
Gene Alias A630086M08Rik; A930036P18Rik; AA408306; AI447644; AI448652; AU015035; CCAP7; CDC5L complex-associated protein 7; CTD-binding SR-like protein rA8; fc50c06; fd99c08; KIAA1116; mKIAA1116; protein SCAF8; putative RNA-binding protein 16; RBM16; RNA binding motif protein 16; RNA binding motif protein 16-like; protein SCAF8; RNA-binding motif protein 16; SCAF8; SR-like CTD-associated factor 8; SR-related and CTD-associated factor 8; SR-related CTD associated factor 8; SR-related CTD-associated factor 8; wu:fc50c06; wu:fd99c08
Gene Symbols Scaf8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCAF8. Recombinant protein control fragment (Product #RP-95057).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22828
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.