Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCAF8 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549488
Description
May play a role in mRNA processing.
Specifications
SCAF8 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9UPN6 | |
SCAF8 | |
KLH conjugated synthetic peptide between 520-554 amino acids from the Central region of human SCAF8. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SCAF8 | |
Protein SCAF8, CDC5L complex-associated protein 7, RNA-binding motif protein 16, SR-related and CTD-associated factor 8, SCAF8, CCAP7, KIAA1116, RBM16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22828 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction