Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCAF8/RBM16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7247250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166600 20 μg
NB166599 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166600 Supplier Novus Biologicals Supplier No. NBP31575220UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCAF8/RBM16 Polyclonal antibody specifically detects SCAF8/RBM16 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen SCAF8/RBM16
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias CCAP7, CDC5L complex-associated protein 7, KIAA1116RNA-binding protein 16, putative RNA-binding protein 16, RBM16, RNA binding motif protein 16, RNA-binding motif protein 16, SR-like CTD-associated factor 8, SR-related and CTD-associated factor 8, SR-related CTD-associated factor 8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 572-634 of human SCAF8/RBM16 (NP_055707.3).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22828
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.