Learn More
Mopec Scalpel Blades, Non-Sterile, 100 pack
Shop All Mopec Products
Click to view available options
Size:
# 22
# 70
Description
- Individually wrapped
- Non-sterile carbon steel
- 100 pack
Specifications
Specifications
| Blade Material | Carbon Steel |
| Product Type | Scalpel Blade |
| Size | # 70 |
| Sterility | Non-sterile |
| Quantity | 10/Pk. |