Learn More
Cincinnati Surgical Sterile-Stainless Steel Blades and Scalpels
Click to view available options
Size:
# 10
# 11
# 12
# 15
# 15C
# 20
# 21
# 22
Quantity:
10/Pk.
25/Pk.
Product Type:
Blade
Safety Scalpel
Description
- Made from British stainless steel
- 200% visual inspection
- Narrow grinding angle resulting in increased initial keenness
- Medium surface finish
- Strong foil package
- Handles are made of plastic
Specifications
Specifications
| Blade Material | British Stainless Steel |
| Product Type | Safety Scalpel |
| Size | # 10 |
| For Use With (Equipment) | Flexible Blade Cover |
| Description | Medium Surface Finish |
| Sterility | Sterile |
| Quantity | 10/Pk. |