Learn More
Invitrogen™ SCAMP2 Monoclonal Antibody (OTI3C2)

Description
SCAMP2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | SCAMP2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI3C2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Scamp2 |
| Gene Accession No. | O15127 |
| Gene Alias | AI875466; I79_023583; Sc2; scamp 2; SCAMP2; Secretory carrier membrane protein 2; secretory carrier-associated membrane protein 2; testis secretory sperm-binding protein Li 219p |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.