Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCAMP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112835
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112835 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112835 Supplier Invitrogen™ Supplier No. PA5112835
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCAMP3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

This gene product belongs to the SCAMP family of proteins which are secretory carrier membrane proteins. They function as carriers to the cell surface in post-golgi recycling pathways. Different family members are highly related products of distinct genes, and are usually expressed together. These findings suggest that the SCAMPs may function at the same site during vesicular transport rather than in separate pathways. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCAMP3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SCAMP3
Gene Accession No. O14828
Gene Alias C1orf3; propin 1; PROPIN1; Sc3; scamp3; Secretory carrier membrane protein 3; secretory carrier membrane protein 3 isoform 1; secretory carrier-associated membrane protein 3; TU52; zgc:103488
Gene Symbols SCAMP3
Host Species Rabbit
Immunogen Recombinant Human Secretory carrier-associated membrane protein 3 protein (2-170AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10067
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.