Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCAMP4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560309
Description
Immunogen sequence: KVHRIYRGAG GSFQKAQTEW NTGTWRNPPS REAQYNNFSG NSLPEYPTVP SYPGSGQWP Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.
SCAMPs, or secretory carrier membrane proteins, are membrane proteins that are widely distributed and play a role in membrane trafficking. Most SCAMPs, such as SCAMP1, have N-terminal cytoplasmic NPF repeats, 4 central transmembrane regions, and a short C-terminal cytoplasmic tail. These NPF repeats are likely involved in mediating endocytosis. However, other SCAMPs, like SCAMP4, lack the NPF repeats and are thus unlikely to function in endocytosis. SCAMP4 has been associated with diseases such as Neurodevelopmental Disorder With Brain Abnormalities, Poor Growth, And Dysmorphic Facies and Strabismus.
Specifications
SCAMP4 | |
Polyclonal | |
Unconjugated | |
SCAMP4 | |
2410022D05Rik; AI448940; AU022392; AW742369; Sc4; scam4; Scamp4; SCAMP-4; Secretory carrier membrane protein 4; secretory carrier-associated membrane protein 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
113178 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969E2 | |
SCAMP4 | |
Recombinant protein corresponding to Human SCAMP4. Recombinant protein control fragment (Product #RP-99248). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction