Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    SCAMP5 Antibody, Allophycocyanin, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1203432APC
Description
SCAMP5 Polyclonal antibody specifically detects SCAMP5 in Rat samples. It is validated for Western BlotSpecifications
| SCAMP5 | |
| Polyclonal | |
| Western Blot | |
| DKFZp686L1799, hSCAMP5, MGC24969, Sc5, secretory carrier membrane protein 5, secretory carrier-associated membrane protein 5 | |
| Synthetic Peptide: C N(217) Q P Q T Q Y S A T P N Y T Y S N(233) | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones | |
| 192683 | |
| Store at 4C in the dark. | |
| IgG | 
| Western Blot | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction