Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCAMP5 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1203432MFV500
Description
SCAMP5 Polyclonal antibody specifically detects SCAMP5 in Rat samples. It is validated for Western BlotSpecifications
SCAMP5 | |
Polyclonal | |
Western Blot | |
DKFZp686L1799, hSCAMP5, MGC24969, Sc5, secretory carrier membrane protein 5, secretory carrier-associated membrane protein 5 | |
Synthetic Peptide: C N(217) Q P Q T Q Y S A T P N Y T Y S N(233) | |
0.1 mL | |
Membrane Trafficking and Chaperones | |
192683 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction