Learn More
Interscience Scan 50
Click to view available options
Includes:
HandPad, Headphones
Description
Scan 50 is a manual colony counter with improved ergonomics that help prevent Musculoskeletal Disorders. It offers contrasting illumination of colonies while protecting the user's eyes. Traceability is ensured by a USB port.
- Counting on: 55 to 90 mm dia. Petri dishes, Petrifilm™, MC-media Pads™, Compact Dry™, EasyPlate™, filtration membranes
- Dark Field 2: Indirect LED lighting
- Data export via USB, LIMS, under CSV format (Excel™) or text format (Word™)
The Scan 50 Pro includes headphones and an ergonomic arm rest.
Specifications
Specifications
Product Type | Darkfield Colony Counter |
Display Type | LED |
Voltage | 100/240 V |