Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCAND1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP256092
Description
SCAND1 Polyclonal specifically detects SCAND1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SCAND1 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
RAZ1, SCAN domain containing 1, SCAN domain-containing 1, SCAN-related protein RAZ1, SDP1SCAN domain-containing protein 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
SCAND1 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PSSPNAAVPEAIPTPRAAASAALELPLGPAPVSVAPQAEAEARSTPGPAGSRLGPETFRQRF | |
100 μL | |
Cell Cycle and Replication | |
51282 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction