Learn More
Invitrogen™ SCAND1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total SCAND1 protein.
Specifications
Specifications
| Antigen | SCAND1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SCAND1 |
| Gene Accession No. | P57086 |
| Gene Alias | 2310003H23Rik; DIPP; DIPP1; Leap1; leucine-rich-domain inter-acting protein 1; peroxisome proliferative activated receptor, gamma, coactivator 2; PGC-2; Ppargc2; RAZ1; SCAN domain containing 1; SCAN domain-containing 1; SCAN domain-containing protein 1; Scand1; SCAN-related protein RAZ1; SDP1 |
| Gene Symbols | SCAND1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.