Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCAND1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104282
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104282 Supplier Invitrogen™ Supplier No. PA5104282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SCAND1.

SCAND1 (SCAN domain containing 1), also known as RAZ1 or SDP1, is a nuclear protein that may play a role in the mechanism of transcription regulation. It is widely expressed with highest levels of expression found in kidney, thyroid, liver, prostate and testis. SCAND1 contains one SCAN box domain and, unlike the majority of other SCAN domain containing proteins, it does not contain a zinc finger motif. The SCAN box domain is a conserved leucine rich motif, approximately 60 amino acids in length, that participates in proteinprotein interactions. SCAND1 interacts with ZNF38, ZNF191 and MZF-1. It also interacts with ZNF202 and PPARgamma, suggesting that it plays a role in the transcriptional regulation of genes that are involved in energy homeostasis and lipid metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCAND1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SCAND1
Gene Accession No. P57086
Gene Alias 2310003H23Rik; DIPP; DIPP1; Leap1; leucine-rich-domain inter-acting protein 1; peroxisome proliferative activated receptor, gamma, coactivator 2; PGC-2; Ppargc2; RAZ1; SCAN domain containing 1; SCAN domain-containing 1; SCAN domain-containing protein 1; Scand1; SCAN-related protein RAZ1; SDP1
Gene Symbols SCAND1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SCAND1(Accession P57086), corresponding to amino acid residues T11-I61.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362252, 51282
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.