Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCAND1 Polyclonal Antibody
SCAND1 Polyclonal Antibody
Supplier: Thermo Scientific PA5110342
Description
SCAND1 Polyclonal Antibody for Western Blot

Specifications
SCAND1 | |
Polyclonal | |
Unconjugated | |
SCAND1 | |
2310003H23Rik; DIPP; DIPP1; Leap1; leucine-rich-domain inter-acting protein 1; peroxisome proliferative activated receptor, gamma, coactivator 2; PGC-2; Ppargc2; RAZ1; SCAN domain containing 1; SCAN domain-containing 1; SCAN domain-containing protein 1; Scand1; SCAN-related protein RAZ1; SDP1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
19018, 362252, 51282 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.68 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P57086 | |
SCAND1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-179 of human SCAND1 (NP_057642.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction