Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCAND3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569496
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569496 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569496 Supplier Invitrogen™ Supplier No. PA569496
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 100%; Horse: 100%; Human: 100%.

ZBED9 gene ontology annotations related to this gene include DNA integration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCAND3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZBED9
Gene Accession No. Q6R2W3
Gene Alias Buster4; dJ1186N24.3; dJ1186N24.3 (novel zinc finger protein); KIAA1925; SCAN domain containing 3; SCAN domain-containing protein 3; SCAND3; transposon-derived Buster4 transposase-like protein; ZBED9; ZFP38-L; Zinc finger BED domain-containing protein 9; zinc finger BED-type containing 9; zinc finger protein 305 pseudogene 2; zinc finger protein 452; zinc finger, BED-type containing 9; ZNF305P2; ZNF452
Gene Symbols ZBED9
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human SCAND3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114821
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.