Learn More
SCAND3 Polyclonal Antibody

Description
SCAND3 Polyclonal Antibody for ICC/IF, IHC
Specifications
Specifications
| Antigen | SCAND3 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZBED9 |
| Gene Accession No. | Q6R2W3 |
| Gene Alias | Buster4; dJ1186N24.3; dJ1186N24.3 (novel zinc finger protein); KIAA1925; SCAN domain containing 3; SCAN domain-containing protein 3; SCAND3; transposon-derived Buster4 transposase-like protein; ZBED9; ZFP38-L; Zinc finger BED domain-containing protein 9; zinc finger BED-type containing 9; zinc finger protein 305 pseudogene 2; zinc finger protein 452; zinc finger, BED-type containing 9; ZNF305P2; ZNF452 |
| Gene Symbols | ZBED9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.