Alfa Aesar™ Scandium(III) chloride, anhydrous, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1867703

Quantity 1g

 View more versions of this product

Catalog No. AA1867703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10361-84-9
    Formula Weight 151.32
    MDL Number MFCD00011221
    Merck Index 14,8392
    Chemical Name or Material Scandium(III) chloride, Anhydrous

    Specifications

    Formula Weight 151.32
    Mesh Size −20 Mesh
    Odor Odorless
    Sensitivity Hygroscopic
    CAS 10361-84-9
    Merck Index 14,8392
    Melting Point 939°C
    Quantity 1g
    TSCA TSCA
    MDL Number MFCD00011221
    Solubility Information Soluble in water; Insoluble in alcohol
    Density 2.39g/mL
    Chemical Name or Material Scandium(III) chloride, Anhydrous
    Assay 99.9%, (REO)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit