Alfa Aesar™ Scandium(III) chloride hexahydrate, REacton™, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1121806

Quantity 5g

 View more versions of this product

Catalog No. AA1121806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20662-14-0
    Formula Weight 259.41 (151.32 Anhydrous)
    MDL Number MFCD00149852
    Merck Index 14,8392
    Chemical Name or Material Scandium(III) chloride hexahydrate
    Grade Reacton™

    Specifications

    Formula Weight 259.41 (151.32 Anhydrous)
    Physical Form Crystalline Aggregates
    MDL Number MFCD00149852
    TSCA TSCA
    CAS 20662-14-0
    Quantity 5g
    Sensitivity Hygroscopic
    Merck Index 14,8392
    Chemical Name or Material Scandium(III) chloride hexahydrate
    Assay 99.9%, (REO)
    Grade Reacton™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit