Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scandium(III) oxide, 99.998% (metals basis), REM 50ppm, Alfa Aesar™, REacton™

Manufacturer:  Alfa Aesar 1091603

Quantity 1g

 View more versions of this product

Catalog No. AA1091603


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12060-08-1
99.998% (metals basis)
137.91
Scandium(III) oxide, REM 50ppm max.
Powder
Sc2O3
MFCD00011223

Specifications

12060-08-1
99.998% (metals basis)
137.91
Odorless
1g
Powder
Sc2O3
MFCD00011223
White
Scandium(III) oxide, REM 50ppm max.

Safety and Handling

SDS & Safety Documents

EINECSNumber : 235-042-0

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit