Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scandium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific™

Catalog No. p-7051158
Click to view available options
Quantity:
2 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14066-05-8
Molecular Formula Cl3O12Sc
Molecular Weight (g/mol) 343.294
MDL Number MFCD00243289
InChI Key UBVKRMCXDCYZQK-UHFFFAOYSA-K
Synonym scandium perchlorate, scandium 3+ perchlorate, acmc-20ajr3, scandium 3+ ion triperchlorate, trihyperchloric acid scandium salt, scandium iii perchlorate solution, scandium 3+ triperchlorate
PubChem CID 11013318
IUPAC Name scandium(3+);triperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Sc+3]

Specifications

Physical Form Liquid
Quantity 2 g
UN Number UN3211
Solubility Information Insoluble in water.
Formula Weight 451.42 (343.30 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Chemical Name or Material Scandium(III) perchlorate
Hazard Category H271-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
EINECSNumber 237-914-6
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.