Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Scandium(III) Triflate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Scandium(III) triflate is an extremely active, efficient, recoverable and reusable acylation catalyst. Its an important catalyst for the Friedel-Crafts acylation, Diels-Alder reactions and other carbon-carbon bond-forming reactions. It also stereochemically catalyzes the radical polymerization of acrylates. Scandium(III) triflate complex of (4'saS,5'saS)-2,6-bis[4'sa-(triisopropylsilyl)oxymethyl-5'sa-phenyl-1'sa,3'sa-oxazolin-2'sa-yl]pyridine has been employed as catalyst for the asymmetric Friedel-Crafts reaction between substituted indoles and methyl (E)-2-oxo-4-aryl-3-butenoates.Scandium(III) triflate was used as a catalyst in:Hydrothiolation reaction of aromatic and aliphatic thiols.Selective two-electron reduction of O2 by ferrocene derivatives.Vinylogous Friedel-Crafts alkylation of indoles and pyrroles in water.Synthesis of +a-cyanoketones.Combination with triethylsilane to reductively open functionalized pyranoside rings. The key steps of synthesis of bullvalone via a stabilized sulfur ylide.
Specifications
Specifications
| CAS | 144026-79-9 |
| Molecular Formula | C3F9O9S3Sc |
| Linear Formula | Sc(SO3CF3)3 |
| MDL Number | MFCD00192433 |
| Quantity | 1 g |
| Synonym | Sc(OTf)3; Scandium(III) trifluoromethanesulfonate; Trifluoromethanesulfonic acid scandium(III) salt |
| Molecular Weight (g/mol) | 492.16 g/mol |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction