Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Scandium(III) Triflate

Catalog No. 4182185G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-821-85G 5 g
41-821-81G 1 g
41-821-8250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 41-821-85G Supplier Sigma Aldrich Supplier No. 4182185G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Scandium(III) triflate is an extremely active, efficient, recoverable and reusable acylation catalyst. Its an important catalyst for the Friedel-Crafts acylation, Diels-Alder reactions and other carbon-carbon bond-forming reactions. It also stereochemically catalyzes the radical polymerization of acrylates. Scandium(III) triflate complex of (4'saS,5'saS)-2,6-bis[4'sa-(triisopropylsilyl)oxymethyl-5'sa-phenyl-1'sa,3'sa-oxazolin-2'sa-yl]pyridine has been employed as catalyst for the asymmetric Friedel-Crafts reaction between substituted indoles and methyl (E)-2-oxo-4-aryl-3-butenoates.Scandium(III) triflate was used as a catalyst in:Hydrothiolation reaction of aromatic and aliphatic thiols.Selective two-electron reduction of O2 by ferrocene derivatives.Vinylogous Friedel-Crafts alkylation of indoles and pyrroles in water.Synthesis of +a-cyanoketones.Combination with triethylsilane to reductively open functionalized pyranoside rings. The key steps of synthesis of bullvalone via a stabilized sulfur ylide.

Specifications

CAS 144026-79-9
Molecular Formula C3F9O9S3Sc
Linear Formula Sc(SO3CF3)3
MDL Number MFCD00192433
Quantity 5 g
Synonym Sc(OTf)3; Scandium(III) trifluoromethanesulfonate; Trifluoromethanesulfonic acid scandium(III) salt
Molecular Weight (g/mol) 492.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.