Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCAP Monoclonal Antibody (1992C245.37.16)
GREENER_CHOICE

Catalog No. PIMA537768
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537768 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537768 Supplier Invitrogen™ Supplier No. MA537768
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SCAP Monoclonal Antibody for Western Blot

The protein encoded by this gene interacts with thyroid hormone receptor in a ligand-dependent manner and enhances thyroid hormone-dependent activation from thyroid response elements. This protein contains a bromodomain and is thought to be a nuclear receptor coactivator. Three alternatively spliced transcript variants that encode distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCAP
Applications Western Blot
Classification Monoclonal
Clone 1992C245.37.16
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SCAP
Gene Accession No. Q12770
Gene Alias 9530044G19; EMBL:AAI31853.1}; KIAA0199; mKIAA0199; PSEC0227; SCAP; scap {ECO:0000312; Sreb cleavage-activating protein; SREBF chaperone; SREBP cleavage activating protein; SREBP cleavage-activating protein; sterol regulatory element binding protein cleavage-activating protein; sterol regulatory element-binding protein cleavage-activating protein
Gene Symbols SCAP
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 49-83 amino acids from the N-terminal region of human SCAP.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22937
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.