Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528982
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528982 Supplier Invitrogen™ Supplier No. PA528982
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2, MCF-7. Predicted reactivity: Mouse (91%), Rat (95%), Pig (97%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene interacts with thyroid hormone receptor in a ligand-dependent manner and enhances thyroid hormone-dependent activation from thyroid response elements. This protein contains a bromodomain and is thought to be a nuclear receptor coactivator. Three alternatively spliced transcript variants that encode distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCAP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SCAP
Gene Accession No. Q12770, Q6GQT6
Gene Alias 9530044G19; EMBL:AAI31853.1}; KIAA0199; mKIAA0199; PSEC0227; SCAP; scap {ECO:0000312; Sreb cleavage-activating protein; SREBF chaperone; SREBP cleavage activating protein; SREBP cleavage-activating protein; sterol regulatory element binding protein cleavage-activating protein; sterol regulatory element-binding protein cleavage-activating protein
Gene Symbols SCAP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human SCAP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22937, 235623
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.