Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552090
Description
Immunogen sequence: EVWDAIEGVL CCSSEEVSSG ITALVFLDKR IVAARLNGSL DFFSLETHTA LSPLQFRGTP GRGSSPASPV YSSSDTVACH LTHTVPCAHQ KPITALKAAA GRLVTGSQDH TLRVFRLED Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 94%.
The protein encoded by this gene interacts with thyroid hormone receptor in a ligand-dependent manner and enhances thyroid hormone-dependent activation from thyroid response elements. This protein contains a bromodomain and is thought to be a nuclear receptor coactivator. Three alternatively spliced transcript variants that encode distinct isoforms have been identified.
Specifications
| SCAP | |
| Polyclonal | |
| Unconjugated | |
| SCAP | |
| 9530044G19; EMBL:AAI31853.1}; KIAA0199; mKIAA0199; PSEC0227; SCAP; scap {ECO:0000312; Sreb cleavage-activating protein; SREBF chaperone; SREBP cleavage activating protein; SREBP cleavage-activating protein; sterol regulatory element binding protein cleavage-activating protein; sterol regulatory element-binding protein cleavage-activating protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22937 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q12770 | |
| SCAP | |
| Recombinant protein corresponding to Human SCAP. Recombinant protein control fragment (Product #RP-102329). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction