Learn More
SCAP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303554AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein with a sterol sensing domain (SSD) and seven WD domains. In the presence of cholesterol, this protein binds to sterol regulatory element binding proteins (SREBPs) and mediates their transport from the ER to the Golgi. The SREBPs are then proteolytically cleaved and regulate sterol biosynthesis.Specifications
SCAP | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q12770 | |
SCAP | |
Between 450 and 500 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SCAP | |
SREBP cleavage-activating protein, sterol regulatory element binding protein cleavage-activating protein, sterol regulatory element-binding protein cleavage-activating protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22937 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.