Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCAP Polyclonal Antibody
SCAP Polyclonal Antibody
Supplier: Thermo Scientific PA5115869
Description
SCAP Polyclonal Antibody for Western Blot, ICC/IF

Specifications
SCAP | |
Polyclonal | |
Unconjugated | |
SCAP | |
9530044G19; EMBL:AAI31853.1}; KIAA0199; mKIAA0199; PSEC0227; SCAP; scap {ECO:0000312; Sreb cleavage-activating protein; SREBF chaperone; SREBP cleavage activating protein; SREBP cleavage-activating protein; sterol regulatory element binding protein cleavage-activating protein; sterol regulatory element-binding protein cleavage-activating protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22937, 235623, 301024 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
A2RRU4, Q12770, Q6GQT6 | |
SCAP | |
A synthesized peptide derived from human SCAP(Accession Q12770), corresponding to amino acid residues K646-G696. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction