Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Watson Gloves Scape Goat Lined Impact Gloves

Catalog No. 17099812
Click to view available options
Size:
Medium
Large
X-Large

Scape Goat Lined Impact Gloves have 3M Thinsulate™ C100 lining for superior warmth in cold conditions. Hard-wearing full-grain goatskin leather offers exceptional strength and dexterity.

  • 3M Thinsulate™ C100 lining
  • Full-grain goatskin leather
  • Heavy duty rubber on back of hand, fingers and wrist
  • Drivers style with keystone thumb
  • Snug-fitting shirred elastic wrist with slip-on style cuff

Specifications

Certifications/Compliance ANSI Puncture 3, ANSI Impact 2
Certifications/Compliance (Europe) EN388
Chemotherapeutic Rated no
Chlorinated No
Construction Cut and Sewn
Description Goatskin Leather Glove
Coating Material Uncoated
Color White, Black
Exterior Finish Smooth
Hand Ambidextrous
Material Goatskin Leather, Thermoplastic Rubber
Product Type Cold-resistant Gloves
Size Medium
For Use With (Application) Winter
Glove Cuff Style Elasticated
Liner Material C100 Thinsulate
Lining Lined
Series Scape Goat
Length (English) 9.5 in.
Length (Metric) 23 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.