Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCAPER Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561195
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561195 Supplier Invitrogen™ Supplier No. PA561195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSPTKTRHPR KIDLRARYWA FLFDNLRRAV DEIYVTCESD QSVVECKEVL MMLDNYVRDF KALIDWIQLQ EKLEKTDAQS RPTSLAWEVK KMSPGR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

CCNA2/CDK2 regulatory protein that transiently maintains CCNA2 in the cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCAPER
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCAPER
Gene Accession No. Q9BY12
Gene Alias C430017I08; D530014O03Rik; KIAA1454; KIAA1454-like protein; LOW QUALITY PROTEIN: S phase cyclin A-associated protein in the endoplasmic reticulum; MSTP063; S phase cyclin A-associated protein in the endoplasmic reticulum; S phase cyclin A-associated protein in the ER; SCAPER; S-phase cyclin A associated protein in the ER; S-phase cyclin A-associated protein in the ER; Zfp291; Zinc finger protein 291; znf291
Gene Symbols SCAPER
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCAPER. Recombinant protein control fragment (Product #RP-98089).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 49855
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.