Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCAPER Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569015
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569015 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569015 Supplier Invitrogen™ Supplier No. PA569015
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

CCNA2/CDK2 regulatory protein that transiently maintains CCNA2 in the cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCAPER
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SCAPER
Gene Accession No. Q9BY12
Gene Alias C430017I08; D530014O03Rik; KIAA1454; KIAA1454-like protein; LOW QUALITY PROTEIN: S phase cyclin A-associated protein in the endoplasmic reticulum; MSTP063; S phase cyclin A-associated protein in the endoplasmic reticulum; S phase cyclin A-associated protein in the ER; SCAPER; S-phase cyclin A associated protein in the ER; S-phase cyclin A-associated protein in the ER; Zfp291; Zinc finger protein 291; znf291
Gene Symbols SCAPER
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human SCAPER.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 49855
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.