Learn More
Invitrogen™ SCARA3 Polyclonal Antibody

Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (95%), Rat (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | SCARA3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.78 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | SCARA3 |
| Gene Accession No. | Q6AZY7 |
| Gene Alias | APC7; C130058N24Rik; cellular stress response gene protein; cellular stress response protein; CSR; CSR1; macrophage scavenger receptor-like 1; MSLR1; MSRL1; Scara3; Scavenger receptor class A member 3; scavenger receptor class A, member 3 |
| Gene Symbols | SCARA3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.