Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCARA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA548058
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548058 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548058 Supplier Invitrogen™ Supplier No. PA548058
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, less than 40% cross-reactivity with recombinant human SCARA5 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Scavenger receptor class A member 5 (SCARA5), also known as TESR, is part of the scavenger receptor superfamily, which is composed of many members with diverse structures, expression patterns, and functions. SCARA5 is a cell-surface receptor that contains that collagen-like domain and N-terminal scavenger receptor cysteine rich domain typical of the class A scavenger receptor subfamily. Unlike other class A receptors, SCARA5 does not endocytose acetylated or oxidized low density lipoprotein. Instead, SCARA5 binds serum ferritin and mediates its endocytosis from the cell surface, indicating SCARA5 may play a role in non-transferrin iron delivery. At least four isoforms of SCARB1 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCARA5
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene SCARA5
Gene Accession No. Q8K299
Gene Alias 4932433F15Rik; 4933425F03Rik; AV278087; NET33; SCARA5; Scavenger receptor class A member 5; scavenger receptor class A, member 5 (putative); scavenger receptor hlg; Tesr; testis expressed scavenger receptor; UNQ2938/PRO28700
Gene Symbols SCARA5
Host Species Sheep
Immunogen CHO-derived recombinant mouse SCARA5 Arg83-Pro491.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71145
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.