Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCARF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115870
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115870 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115870 Supplier Invitrogen™ Supplier No. PA5115870
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SCARF1.

The protein encoded by this gene is a scavenger receptor that is expressed in endothelial cells and mediates the binding and degradation of acetylated low density lipoprotein (Ac-LDL). Transcription factors ZNF444/EZF-2 and SP1 are found to be responsible for the regulation of this gene. Five alternatively spliced transcript variants encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCARF1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Scarf1
Gene Accession No. Q14162, Q5ND28
Gene Alias AA986099; acetyl LDL receptor; endothelial cells scavenger receptor; KIAA0149; MGC47738; mKIAA0149; SCARF1; Scavenger receptor class F member 1; scavenger receptor class F, member 1; scavenger receptor expressed by endothelial cells 1; Scavenger receptor expressed by endothelial cells 1 protein; SREC; SREC1; SREC-I
Gene Symbols Scarf1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SCARF1(Accession Q14162), corresponding to amino acid residues G426-S476.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303313, 380713, 8578
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.